Order Entry
United States
Orders LinkContactUsLinkComponent
 

SINGLET™ pH Buffer, Single-Use

±0.02, 25 °C, 20×20 ml pouches, 4.01
Catalog # 104086-650
Marketsource
Supplier:   Hach
 :  2770020
SINGLET™ pH Buffer, Single-Use
Catalog # 104086-650
Supplier:  Hach
Marketsource
CAS Number:  
SINGLET™ pH Buffer, Single-Use
SINGLET™ pH Buffer, Single-Use

±0.02, 25 °C, 20×20 ml pouches, 4.01

CAS Number:  
Price
$74.14
Log in for availability
Pack of 20
 
Product Details & Documents

These buffers solutions packs are used for pH meter calibration. The sealed foil pouches can be transported easily and safely without any spillage and also prevents contamination or exposure to air of the solutions.

  • Ideal for on-site analyses
  • Easy to use
  • Accuracy pH: ±0.02 at 25 °C
  • Traceable to NIST

 

Specifications

Specifications

  • Packed:
    20×20 ml pouches
  • Reference temperature:
    25 °C
  • pH:
    4.01
  • pH accuracy:
    ±0.02

Specifications