Order Entry
United States
Orders LinkContactUsLinkComponent
SINGLET™ pH Buffer, Single-Use

SINGLET™ pH Buffer, Single-Use

20/pack (10 each of 4.01 and 7.00 pH), 4.01/7.00
Catalog # 104086-648
Marketsource
Supplier:   Hach
Supplier #:  2769920
SINGLET™ pH Buffer, Single-Use
Catalog # 104086-648
Supplier:  Hach
Marketsource
CAS Number:  
SINGLET™ pH Buffer, Single-Use
SINGLET™ pH Buffer, Single-Use

20/pack (10 each of 4.01 and 7.00 pH), 4.01/7.00

CAS Number:  
Price
$72.95
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

These buffers solutions packs are used for pH meter calibration. The sealed foil pouches can be transported easily and safely without any spillage and also prevents contamination or exposure to air of the solutions.

  • Ideal for on-site analyses
  • Easy to use
  • Accuracy pH: ±0.02 at 25 °C
  • Traceable to NIST

Documents

Specifications

Specifications

  • Packed:
    20/pack (10 each of 4.01 and 7.00 pH)
  • pH:
    4.01/7.00

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.