Order Entry
United States
Orders LinkContactUsLinkComponent
SINGLET™ pH Buffer, Single-Use

SINGLET™ pH Buffer, Single-Use

20/pack (10 each of 7.00 and 10.01 pH), 7.00/10.01
Catalog # 104086-646
Marketsource
Supplier:   Hach
Supplier #:  2769820
SINGLET™ pH Buffer, Single-Use
Catalog # 104086-646
Supplier:  Hach
Marketsource
CAS Number:  
SINGLET™ pH Buffer, Single-Use
SINGLET™ pH Buffer, Single-Use

20/pack (10 each of 7.00 and 10.01 pH), 7.00/10.01

CAS Number:  
Price
$72.95
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

These buffers solutions packs are used for pH meter calibration. The sealed foil pouches can be transported easily and safely without any spillage and also prevents contamination or exposure to air of the solutions.

  • Ideal for on-site analyses
  • Easy to use
  • Accuracy pH: ±0.02 at 25 °C
  • Traceable to NIST

Documents

Specifications

Specifications

  • Packed:
    20/pack (10 each of 7.00 and 10.01 pH)
  • pH:
    7.00/10.01

Specifications