Order Entry
United States
Orders LinkContactUsLinkComponent
pH Buffer Solutions, Color Coded

pH Buffer Solutions, Color Coded

±0.02, Phthalate, 25 °C, 4.01
  104086-054
Marketsource
 :   Hach
Supplier #:  2283426
pH Buffer Solutions, Color Coded
  104086-054
 :  Hach
Marketsource
 :  
pH Buffer Solutions, Color Coded
pH Buffer Solutions, Color Coded

±0.02, Phthalate, 25 °C, 4.01

 :  
 
$19.09
   
Each
Minimum order quantity is 1
 
  • For calibration of pH meters
  • Color coded for easy identification
  • Accuracy: pH is ±0.02 at 25 °C

Certifications: Traceable to NIST.

Documents

 

 

  • pH buffer type:
    Phthalate
  • Reference temperature:
    25 °C
  • pH:
    4.01
  • pH accuracy:
    ±0.02

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.