Buffer Solution for Chlorine Determination
Phosphate, 7
Catalog # 104085-876
Marketsource
Supplier: Hach
Supplier #: 2155332
Buffer Solution for Chlorine Determination
Catalog # 104085-876
Supplier: Hach
Marketsource
CAS Number:
Jump to:
Product Details & Documents
For determination of chlorine by amperometric titration. APHA formulation 100 ml bottle.
Specifications
Specifications
- pH buffer type:Phosphate
- pH:7



