Orders LinkContactUsLinkComponent
TPH Reagent Sets
TPH Reagent Sets
Triazole * Only for the DR 5000 UV-Vis, UV photolysis, 1 - 20 mg/L, -, -, PP
Catalog # 104085-864
Marketsource
Supplier:   Hach
Supplier #:  2141299
TPH Reagent Sets
Catalog # 104085-864
Supplier:  Hach
Marketsource
CAS Number:  
TPH Reagent Sets
TPH Reagent Sets
Triazole * Only for the DR 5000 UV-Vis, UV photolysis, 1 - 20 mg/L, -, -, PP
CAS Number:  
Price
$129.74
Log in for availability
Pack of 100
Minimum order quantity is 1
Product Details & Documents
For the determination of total petroleum hydrocarbons (TPH) in soil and water.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.
Documents
Specifications
Specifications
  • Chemical method:
    UV photolysis
  • Description:
    Triazole * Only for the DR 5000 UV-Vis
  • Range:
    1 - 20 mg/L
  • Type:
    PP
  • Sample Size:
  • No. of tests:
Specifications