Order Entry
United States
Orders LinkContactUsLinkComponent
pH Test Kits, Model 17

pH Test Kits, Model 17

pH, mid range, model 17J, Thymol blue, pH 7.4 - 9.6
Catalog # 104085-534
Marketsource
Supplier:   Hach
Supplier #:  147009
pH Test Kits, Model 17
Catalog # 104085-534
Supplier:  Hach
Marketsource
CAS Number:  
pH Test Kits, Model 17
pH Test Kits, Model 17

pH, mid range, model 17J, Thymol blue, pH 7.4 - 9.6

CAS Number:  
Price
$216.00
Log in for availability
Pack of 100
Minimum order quantity is 1
Product Details & Documents

It is a simple, low cost colour disc method which is one of the most accurate visual test kit methods for pH.

Delivery information: Kit includes indicator reagent, colour disc, comparator box, two viewing tubes, instructions and carrying case.

Specifications

Specifications

  • Chemical method:
    Thymol blue
  • Description:
    pH, mid range, model 17J
  • Range:
    pH 7.4 - 9.6

Specifications