Orders LinkContactUsLinkComponent
Test Kits, Drop Count Tests
Chloride, model 8-P, use silver nitrate titrant, 5 - 100 mg/L Cl-; 20 - 400 mg/L Cl-
Catalog # 104085-456
Marketsource
Supplier:   Hach
 :  144001
Test Kits, Drop Count Tests
Catalog # 104085-456
Supplier:  Hach
Marketsource
CAS Number:  
Test Kits, Drop Count Tests
Test Kits, Drop Count Tests
Chloride, model 8-P, use silver nitrate titrant, 5 - 100 mg/L Cl-; 20 - 400 mg/L Cl-
CAS Number:  
Price
$138.45
Log in for availability
Each
Product Details & Documents
The drop count test is a simple, easy and cost-effective way to test for many parameters like chlorine, chloride, dissolved oxygen, carbon dioxide, sulphite, nitrogen, formaldehyde, and more.
  • Portable - ideal for use in field or in lab
  • Used as a quick reference or for confirmatory test with color indicator
  • Reagents supplied with the kit
 : Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifications
Specifications
  • Description:
    Chloride, model 8-P, use silver nitrate titrant
  • Range:
    5 - 100 mg/L Cl-; 20 - 400 mg/L Cl-
Specifications