Orders LinkContactUsLinkComponent
Test Kits, Drop Count Tests
Chloride reagent set for 8-P, drop count titration, set includes chloride 2 indicator powder pillows and silver nitrate titrant (0.0493 N titrant), -
  104086-264
Marketsource
 :   Hach
 :  2437600
Test Kits, Drop Count Tests
  104086-264
 :  Hach
Marketsource
 :  
Test Kits, Drop Count Tests
Test Kits, Drop Count Tests
Chloride reagent set for 8-P, drop count titration, set includes chloride 2 indicator powder pillows and silver nitrate titrant (0.0493 N titrant), -
 :  
$117.89
Each
The drop count test is a simple, easy and cost-effective way to test for many parameters like chlorine, chloride, dissolved oxygen, carbon dioxide, sulphite, nitrogen, formaldehyde, and more.
  • Portable - ideal for use in field or in lab
  • Used as a quick reference or for confirmatory test with color indicator
  • Reagents supplied with the kit
 : Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
  • Description:
    Chloride reagent set for 8-P, drop count titration, set includes chloride 2 indicator powder pillows and silver nitrate titrant (0.0493 N titrant)
  • Range:
Recommendations