BUFFER SOLU BLU PH10.01 500ML
Catalog # 104086-076
Supplier: Hach
Frequently Bought Together
Whirl-Pak® Sterilized Stand-Up Bags, Nasco Sampling
Supplier: NASCO SAMPLING LLC
VWR® Boston Round Bottles, Narrow Mouth, Clear and Amber
Supplier: VWR International
ep Dualfilter T.I.P.S.® Racks, Filter Pipette Tips
Supplier: Eppendorf
Accessories for Tissue-Tek® Film® Coverslipper, Sakura
Supplier: Sakura Finetek
CrossLab CE and CE/MS Filters and Frits, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
KIMAX® Berzelius Beakers, Tall Form, Graduated, Borosilicate Glass, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
OPTICLEAR™ Vials, Borosilicate Glass, with Polyethylene Stopper, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Sodium chloride solution 0.9% USP, sterile (Normal saline)
Supplier: Cytiva
VWR® Redi-Stor™ Conductivity Probe Storage Solution
Supplier: VWR International
Standard TouchMaster Cotton Lisle Inspection Gloves, Magid
Supplier: Magid Glove
Captiva Syringe Filters, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
HYDROCHLORIC ACID 5N SOL
Supplier: Aqua Solutions
BUFFER SOLU YLW PH7.00 500ML
Supplier: Hach
BUFFER SOLU RED PH4.01 500ML
Supplier: Hach
Aniline Blue
Supplier: Ward's Science
Buffers, Reference Standards, Color-Coded, Ricca Chemical Company
Supplier: Ricca Chemical
Columbia CNA Agar with Sheep Blood, Hardy Diagnostics
Supplier: Hardy Diagnostics
Accessories for Akro-Grid/Nest and Stack Totes, Akro-Mils
Supplier: Akro-Mils