Order Entry
United States
Orders LinkContactUsLinkComponent
 

pH Buffer Solutions, Color Coded

±0.02, Phosphate, 25 °C, 7
Catalog # 104086-072
Marketsource
Supplier:   Hach
 :  2283561
pH Buffer Solutions, Color Coded
Catalog # 104086-072
Supplier:  Hach
Marketsource
CAS Number:  
pH Buffer Solutions, Color Coded
pH Buffer Solutions, Color Coded

±0.02, Phosphate, 25 °C, 7

CAS Number:  
Price
$300.46
Log in for availability
Each
 
Product Details & Documents
  • For calibration of pH meters
  • Color coded for easy identification
  • Accuracy: pH is ±0.02 at 25 °C

 : Traceable to NIST.

 

Specifications

Specifications

  • pH buffer type:
    Phosphate
  • Reference temperature:
    25 °C
  • pH:
    7
  • pH accuracy:
    ±0.02

Specifications