Order Entry
United States
Orders LinkContactUsLinkComponent
 

pH Buffer Solutions, Color Coded

±0.02, Phosphate, 25 °C, 7
Catalog # 104086-066
Marketsource
Supplier:   Hach
 :  2283536
pH Buffer Solutions, Color Coded
Catalog # 104086-066
Supplier:  Hach
Marketsource
CAS Number:  
pH Buffer Solutions, Color Coded
pH Buffer Solutions, Color Coded

±0.02, Phosphate, 25 °C, 7

CAS Number:  
Price
$12.45
Log in for availability
Each
 
Product Details & Documents
  • For calibration of pH meters
  • Color coded for easy identification
  • Accuracy: pH is ±0.02 at 25 °C

Certifications: Traceable to NIST.

Documents

Specifications

Specifications

  • pH buffer type:
    Phosphate
  • Reference temperature:
    25 °C
  • pH:
    7
  • pH accuracy:
    ±0.02

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.