Order Entry
United States
Orders LinkContactUsLinkComponent
pH Buffer Solutions, Color Coded

pH Buffer Solutions, Color Coded

±0.02, Phosphate, 25 °C, 7
Catalog # 104086-064
Marketsource
Supplier:   Hach
Supplier #:  2283526
pH Buffer Solutions, Color Coded
Catalog # 104086-064
Supplier:  Hach
Marketsource
CAS Number:  
pH Buffer Solutions, Color Coded
pH Buffer Solutions, Color Coded

±0.02, Phosphate, 25 °C, 7

CAS Number:  
Price
$19.09
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
  • For calibration of pH meters
  • Color coded for easy identification
  • Accuracy: pH is ±0.02 at 25 °C

Certifications: Traceable to NIST.

Documents

Specifications

Specifications

  • pH buffer type:
    Phosphate
  • Reference temperature:
    25 °C
  • pH:
    7
  • pH accuracy:
    ±0.02

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.