Order Entry
United States
Orders LinkContactUsLinkComponent
pH Buffer Solutions, Color Coded

pH Buffer Solutions, Color Coded

±0.02, Phthalate, 25 °C, 4.01
Catalog # 104086-062
Marketsource
Supplier:   Hach
Supplier #:  2283461
pH Buffer Solutions, Color Coded
Catalog # 104086-062
Supplier:  Hach
Marketsource
CAS Number:  
pH Buffer Solutions, Color Coded
pH Buffer Solutions, Color Coded

±0.02, Phthalate, 25 °C, 4.01

CAS Number:  
Price
$300.46
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
  • For calibration of pH meters
  • Color coded for easy identification
  • Accuracy: pH is ±0.02 at 25 °C

Certifications: Traceable to NIST.

Documents

Specifications

Specifications

  • pH buffer type:
    Phthalate
  • Reference temperature:
    25 °C
  • pH:
    4.01
  • pH accuracy:
    ±0.02

Specifications