Orders LinkContactUsLinkComponent
Test Kits, Drop Count Tests
Test Kits, Drop Count Tests
Chloride, model CD-51, 500 - 10000 mg/L Cl-; 5000 - 100000 mg/L Cl-
Catalog # 104085-752
Marketsource
Supplier:   Hach
Supplier #:  208601
Test Kits, Drop Count Tests
Catalog # 104085-752
Supplier:  Hach
Marketsource
CAS Number:  
Test Kits, Drop Count Tests
Test Kits, Drop Count Tests
Chloride, model CD-51, 500 - 10000 mg/L Cl-; 5000 - 100000 mg/L Cl-
CAS Number:  
Price
$301.85
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
The drop count test is a simple, easy and cost-effective way to test for many parameters like chlorine, chloride, dissolved oxygen, carbon dioxide, sulphite, nitrogen, formaldehyde, and more.
  • Portable - ideal for use in field or in lab
  • Used as a quick reference or for confirmatory test with color indicator
  • Reagents supplied with the kit
Caution: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifications
Specifications
  • Description:
    Chloride, model CD-51
  • Range:
    500 - 10000 mg/L Cl-; 5000 - 100000 mg/L Cl-