Order Entry
United States
Orders LinkContactUsLinkComponent
pH Indicator solution, wide range pH 4-10

pH Indicator solution, wide range pH 4-10

Catalog # 104086-120
Flammable
Irritant
Marketsource
Supplier:   Hach
Supplier #:  2329332
pH Indicator solution, wide range pH 4-10
Catalog # 104086-120
Supplier:  Hach
Marketsource
CAS Number:  
pH Indicator solution, wide range pH 4-10
pH Indicator solution, wide range pH 4-10
CAS Number:  
Price
$46.23
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

For visual determination of pH, using test kits

Method: 10222

  • UN:1219
  • ADR:3,II

Documents

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.