Orders LinkContactUsLinkComponent
Test Kits, Drop Count Tests
Test Kits, Drop Count Tests
Hydrogen peroxide, model HYP-1, 1 - 10 mg/L
Catalog # 104086-084
Marketsource
Supplier:   Hach
Supplier #:  2291700
Test Kits, Drop Count Tests
Catalog # 104086-084
Supplier:  Hach
Marketsource
CAS Number:  
Test Kits, Drop Count Tests
Test Kits, Drop Count Tests
Hydrogen peroxide, model HYP-1, 1 - 10 mg/L
CAS Number:  
Price
$209.08
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
The drop count test is a simple, easy and cost-effective way to test for many parameters like chlorine, chloride, dissolved oxygen, carbon dioxide, sulphite, nitrogen, formaldehyde, and more.
  • Portable - ideal for use in field or in lab
  • Used as a quick reference or for confirmatory test with color indicator
  • Reagents supplied with the kit
Caution: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifications
Specifications
  • Description:
    Hydrogen peroxide, model HYP-1
  • Range:
    1 - 10 mg/L
Specifications