Buffer Solution for Chlorine Determination
Acetate, 4
Catalog # 104085-580
Marketsource
Supplier: Hach
Supplier #: 1490932
Buffer Solution for Chlorine Determination
Catalog # 104085-580
Supplier: Hach
Marketsource
CAS Number:
Jump to:
Product Details & Documents
For determination of chlorine by amperometric titration. APHA formulation 100 ml bottle.
Specifications
Specifications
- pH buffer type:Acetate
- pH:4
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



