Order Entry
United States
Orders LinkContactUsLinkComponent
Buffer Powder Pillows

Buffer Powder Pillows

±0.02, Borate, 25 °C, Pack of 50, 9, Colorless
Catalog # 104085-338
Marketsource
Supplier:   Hach
Supplier #:  1410766
Buffer Powder Pillows
Catalog # 104085-338
Supplier:  Hach
Marketsource
CAS Number:  
Buffer Powder Pillows
Buffer Powder Pillows

±0.02, Borate, 25 °C, Pack of 50, 9, Colorless

CAS Number:  
Price
$54.20
Log in for availability
Pack of 50
Minimum order quantity is 1
Product Details & Documents

The unit-dose powder pillows assure buffer freshness and eliminate the possibility of contamination. Each powder pillow prepares 50 ml of buffer solutions of respective pH.

  • Fresh and reliable
  • No danger of contamination
  • Traceable to NIST standards

Delivery information: The buffer pillows are delivered in an airtight container and are supplied with a certificate of conformity.

Specifications

Specifications

  • Color:
    Colorless
  • Packed:
    Pack of 50
  • pH buffer type:
    Borate
  • Reference temperature:
    25 °C
  • pH:
    9
  • pH accuracy:
    ±0.02

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.