Order Entry
United States
Orders LinkContactUsLinkComponent
 

Buffer Powder Pillows

±0.02, Phosphate, 25 °C, Pack of 15, 6.86, Colorless
Catalog # 104085-334
Marketsource
Supplier:   Hach
 :  1409895
Buffer Powder Pillows
Catalog # 104085-334
Supplier:  Hach
Marketsource
CAS Number:  
Buffer Powder Pillows
Buffer Powder Pillows

±0.02, Phosphate, 25 °C, Pack of 15, 6.86, Colorless

CAS Number:  
Price
$31.28
Log in for availability
Pack of 15
 
Product Details & Documents

The unit-dose powder pillows assure buffer freshness and eliminate the possibility of contamination. Each powder pillow prepares 50 ml of buffer solutions of respective pH.

  • Fresh and reliable
  • No danger of contamination
  • Traceable to NIST standards

 : The buffer pillows are delivered in an airtight container and are supplied with a certificate of conformity.

Specifications

Specifications

  • Color:
    Colorless
  • Packed:
    Pack of 15
  • pH buffer type:
    Phosphate
  • Reference temperature:
    25 °C
  • pH:
    6.86
  • pH accuracy:
    ±0.02

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.