Order Entry
United States
Orders LinkContactUsLinkComponent
pH Buffer Solutions

pH Buffer Solutions

4.01
Catalog # 104085-252
Marketsource
Supplier:   Hach
Supplier #:  1222349
pH Buffer Solutions
Catalog # 104085-252
Supplier:  Hach
Marketsource
CAS Number:  
pH Buffer Solutions
pH Buffer Solutions

4.01

CAS Number:  
Price
$28.17
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

For calibration of pH meters.

Accuracy: pH is ±0.02 at 25 °C

Certifications: Traceable to NIST.

Caution: Please note that the pH 4 and 10 buffers contain formaldehyde <0.1% and the pH 7 buffer contains formaldehyde 0.1 to 1%.

Specifications

Specifications

  • pH:
    4.01

Specifications