Order Entry
United States
Orders LinkContactUsLinkComponent
BUFFER SOLU PH7.00 4L

BUFFER SOLU PH7.00 4L

Catalog # 104085-250
Marketsource
Supplier:   Hach
Supplier #:  1222256
BUFFER SOLU PH7.00 4L
Catalog # 104085-250
Supplier:  Hach
Marketsource
CAS Number:  
BUFFER SOLU PH7.00 4L
CAS Number:  
Price
$108.83
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents