Order Entry
United States
Orders LinkContactUsLinkComponent
pH Buffer Solutions

pH Buffer Solutions

7.00
Catalog # 104085-248
Marketsource
Supplier:   Hach
Supplier #:  1222249
pH Buffer Solutions
Catalog # 104085-248
Supplier:  Hach
Marketsource
CAS Number:  
pH Buffer Solutions
pH Buffer Solutions

7.00

CAS Number:  
Price
$28.17
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

For calibration of pH meters.

Accuracy: pH is ±0.02 at 25 °C

Certifications: Traceable to NIST.

Caution: Please note that the pH 4 and 10 buffers contain formaldehyde <0.1% and the pH 7 buffer contains formaldehyde 0.1 to 1%.

Specifications

Specifications

  • pH:
    7.00

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.