Orders LinkContactUsLinkComponent
pH Buffer Solutions
10
  104085-242
Marketsource
 :   Hach
 :  1222149
pH Buffer Solutions
  104085-242
 :  Hach
Marketsource
 :  
pH Buffer Solutions
pH Buffer Solutions
10
 :  
$28.17
Each
For calibration of pH meters.
Accuracy: pH is ±0.02 at 25 °C
 : Traceable to NIST.
 : Please note that the pH 4 and 10 buffers contain formaldehyde <0.1% and the pH 7 buffer contains formaldehyde 0.1 to 1%.
  • pH:
    10