Orders LinkContactUsLinkComponent
TPH Reagent Sets
TPH Reagent Sets
Iron, FerroVer®, 1,10-Phenanthroline, 0.02 - 3 mg/L, -, 10 ml, PP
Catalog # 104085-778
Marketsource
Supplier:   Hach
Supplier #:  2105728
TPH Reagent Sets
Catalog # 104085-778
Supplier:  Hach
Marketsource
CAS Number:  
TPH Reagent Sets
TPH Reagent Sets
Iron, FerroVer®, 1,10-Phenanthroline, 0.02 - 3 mg/L, -, 10 ml, PP
CAS Number:  
Price
$546.92
Log in for availability
Pack of 1,000
Minimum order quantity is 1
Product Details & Documents
For the determination of total petroleum hydrocarbons (TPH) in soil and water.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.
Documents
Specifications
Specifications
  • Chemical method:
    1,10-Phenanthroline
  • Description:
    Iron, FerroVer®
  • Range:
    0.02 - 3 mg/L
  • Type:
    PP
  • Sample Size:
    10 ml
  • No. of tests:
Specifications