Valves
Valves for CAVRO* XE, M6 Fitting,1/4"-28 UNF, 3-Port-120° flow path, PTFE/PCTFE
Catalog # 77506-582
Supplier: INNOVATIVE LABOR SYSTEME GMBH
Supplier #: 2700080
Jump to:
Product Details & Documents
Specifications
- Description:Valves for CAVRO* XE, M6 Fitting,1/4"-28 UNF, 3-Port-120° flow path, PTFE/PCTFE