Carson MicroMini™ Pocket Microscope
MicroMini™ pocket microscope, Blue
Catalog # 470355-722
Supplier: CARSON OPTICAL
Supplier #: MM-280B
Jump to:
Product Details & Documents
Explore anything…anywhere!
- Uses 3 G12 batteries (included)
- Compact, portable and easy to use
- Attached keychain
The MicroMini™ is a 20×, LED lighted pocket microscope that is always at the ready! It is compact, portable and easy to use. It is so small it can fit in the palm of your hand. Clip on to any hand bag or backpack using the attached key chain. View fabric, flowers, foliage, printing pixels and more all up close! View the hidden objects in currency with the included UV light.
Documents
Specifications
- Magnification:20×
- Color:Blue
- Description:MicroMini™ pocket microscope