Order Entry
United States
Orders LinkContactUsLinkComponent
SoNa™ 520

SoNa™ 520

10x50 µg
Catalog # 77461-862
Supplier:   AAT Bioquest
Supplier #:  21321
SoNa™ 520
Catalog # 77461-862
Supplier:  AAT Bioquest
CAS Number:  
SoNa™ 520
SoNa™ 520

10x50 µg

CAS Number:  
Price
$322.48
Log in for availability
Each(1/Itm/Bx)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

SoNa™ 520 is a new sodium-sensitive fluorescent indicator dye used to detect sodium levels in cells and other biological samples.

  • Stable for 12 months

Quality tested in-house

Caution: For Research Use Only

  • Molecular Weight:720.72 g/mol
Specifications

Specifications

  • Size:
    10x50 µg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.