Orders LinkContactUsLinkComponent
TPH Reagent Sets
MBTH powder pillows, 50 - 100 tests, -, 0.003 - 0.5 mg/L, -, -, PP
Catalog # MSPP-2257169
Marketsource
Supplier:   HACH COMPANY MS
 :  2257169
TPH Reagent Sets
Catalog # MSPP-2257169
Supplier:  HACH COMPANY MS
Marketsource
CAS Number:  
TPH Reagent Sets
TPH Reagent Sets
MBTH powder pillows, 50 - 100 tests, -, 0.003 - 0.5 mg/L, -, -, PP
CAS Number:  
Price
Inquire For Price
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
For the determination of total petroleum hydrocarbons (TPH) in soil and water.
 : Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accommodate these items.
Specifications
Specifications
  • Chemical method:
    -
  • Description:
    MBTH powder pillows, 50 - 100 tests
  • Range:
    0.003 - 0.5 mg/L
  • Type:
    PP
  • Sample Size:
  • No. of tests:
Specifications