ANTIBODY ARC GUINEA PIG PAB RAT 100UL
Catalog # MSPP-156005
Supplier: Cedar Lane
ANTIBODY ARC GUINEA PIG PAB RAT 100UL
Catalog # MSPP-156005
Supplier: Cedar Lane
Supplier Number: 156005
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
Parafilm® M Sealing Film
Supplier: Bemis Company, Inc.
VWR® Weighing Paper
Supplier: VWR International
CiDehol® ST Sterile 70% IPA with WFI, Decon Labs
Supplier: Decon Labs
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Disposable Transfer Pipettes
Supplier: VWR International
VWR® Sterile 70% Isopropanol
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Acetonitrile ≥99.8% for HPLC, Supelco®
Supplier: MilliporeSigma
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
ANTIBODY RECOM NPAS4 MONOCLNAL RAB 100UL
Supplier: ACTIVITY SIGNALING LLC MS