10% TWEEN-20 200MLDNASE/RNASE TEST
Catalog # 77442-666
Supplier: Teknova
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Ethanol absolute ≥100% (v/v) USP for molecular biology (200 Proof)
Supplier: Decon Labs
VWR® Disposable Serological Pipettes
Supplier: VWR International
Parafilm® M Sealing Film
Supplier: Bemis Company, Inc.
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
VWR® Sleeve Stoppers
Supplier: VWR International
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
High Sensitivity DNA ScreenTape Analysis
Supplier: Agilent Technologies
Tris HCl Buffer Solution, Quality Biological
Supplier: QUALITY BIOLOGICAL, INC.
SYBR® Safe DNA Stain, 10,000 X, Edvotek
Supplier: Edvotek
Accessories for Stirling Ultracold Model SU780XLE Ultra-Low Temperature Upright Freezers, Global Cooling
Supplier: Stirling Ultracold
RNA ScreenTape Analysis
Supplier: Agilent Technologies
Quantum™ Simply Cellular® Microspheres, Bangs Laboratories
Supplier: Bangs Laboratories