Order Entry
United States
Orders LinkContactUsLinkComponent
BUFFER CCMB80 500ML

BUFFER CCMB80 500ML

Catalog # 77442-778
Supplier:   Teknova
Supplier #:  C3132
BUFFER CCMB80 500ML
Catalog # 77442-778
Supplier:  Teknova
CAS Number:  
BUFFER CCMB80 500ML
CAS Number:  
Price
$66.00
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.