CARTRIDGE CASCADE REAGENT 5-0.05ML PK10
Catalog # MSPP-RCRC3005
Supplier: CHARLES RIVER ENDOSAFE MS
CARTRIDGE CASCADE REAGENT 5-0.05ML PK10
Catalog # MSPP-RCRC3005
Supplier: CHARLES RIVER ENDOSAFE MS
Supplier Number: RCRC3005
Frequently Bought Together
CrossLab CE and CE/MS Filters and Frits, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
VWR® 11 mm Snap Ring Vials, Kits, Caps, and Inserts, Wide Opening
Supplier: VWR International
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
Screw Top Vials and Caps, 2 ml, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Supplier: VWR International
Closures for 2 ml Screw Top Vials, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Nitrile Examination Gloves, Black
Supplier: VWR International
VWR® Ion Chromatography (IC) PolyVials
Supplier: VWR International
VWR® Screw Caps for 9 mm Screw-Thread Vials, Wide Opening
Supplier: VWR International
Biological Buffers, Teknova
Supplier: Teknova
Mini Dialysis Kit, Cytiva
Supplier: Cytiva