BIOTINYLATED HUMAN HLA-A HLCH82E5200UG
Catalog # 104046-610
Supplier: ACROBIOSYSTEMS
BIOTINYLATED HUMAN HLA-A HLCH82E5200UG
Catalog # 104046-610
Supplier: ACROBIOSYSTEMS
Supplier Number: HLCH82E5200UG
Frequently Bought Together
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Razor Blades
Supplier: VWR International
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Sol-M® Hypodermic Needles
Supplier: SOL-MILLENNIUM MEDICAL
VWR® Chemical-Resistant Laboratory Marker
Supplier: VWR International