ASSAY HICOS JURKAT LUCIA 3-7X10E6-CELL
Catalog # MSPP-JKTL-ICOS
Supplier: Invivogen
ASSAY HICOS JURKAT LUCIA 3-7X10E6-CELL
Catalog # MSPP-JKTL-ICOS
Supplier: Invivogen
Supplier Number: JKTL-ICOS
Frequently Bought Together
Methanol ≥99.8%
Supplier: TCI America
CiDehol® ST Sterile 70% IPA with WFI, Decon Labs
Supplier: Decon Labs
Hexane (mixture of isomers) ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
Accessories for Aspirating Pipettes, Costar®, Corning
Supplier: Corning
Masterflex® L/S® 2-Stop Pump Tubing, Tygon® S3™ E-LFL, Avantor®
Supplier: VWR International
Budget Antibiotic Disks
Supplier: Ward's Science
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Talon® Lab Frame Feet
Supplier: VWR International
Labels, Diversified Biotech
Supplier: Diversified Biotech
High Sensitivity RNA ScreenTape Analysis
Supplier: Agilent Technologies
Using CRISPR to treat Cystic Fibrosis
Supplier: Edvotek
MAGNA AMP LOG 6 PK50
Supplier: MESA LABS MS
CappWash™ Microplate Washer, Pipette.com
Supplier: FIVE RINGS PARTNERS INC MS
High Sensitivity DNA ScreenTape Analysis
Supplier: Agilent Technologies
BUFFER SOLU BLU PH10.01 500ML
Supplier: Hach
VWR® Vinyl Nitrile Hybrid Examination Gloves
Supplier: VWR International