ASSAY UPLEX NHP MCP1 S K156UGK-1
Catalog # 104040-672
Supplier: Meso Scale Diagnostics
ASSAY UPLEX NHP MCP1 S K156UGK-1
Catalog # 104040-672
Supplier: Meso Scale Diagnostics
Supplier Number: K156UGK-1
Frequently Bought Together
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Supplier: Corning
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
Supplier: Corning
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
Clorox® Concentrated Germicidal Bleach
Supplier: Janitorial Supplies
KIMTECH SCIENCE® KIMWIPES™ Delicate Task Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
VWR® Universal Pipette Tips
Supplier: VWR International
Blasticidin S hydrochloride solution 10 mg/ml in HEPES ≥95% (by HPLC)
Supplier: Invivogen
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15×100 mm plate
Supplier: Hardy Diagnostics
DY266-05 HIP-10 DUOSET (1 KT)
Supplier: R&D Systems