ELISA UPLEX TGF COMBO HU SEC K15241K-4
104037-068
Frequently Bought Together
Corning® Centrifuge Tubes, 15 ml
: Corning
Parafilm® M Sealing Film
: Bemis Company, Inc.
VWR® Weighing Paper
: VWR International
VWR® Absorbent Underpads with Waterproof Moisture Barrier
: VWR International
CiDehol® ST Sterile 70% IPA with WFI, Decon Labs
: Decon Labs
Masterflex® L/S® Precision Pump Tubing, Platinum-Cured Silicone, Avantor®
: VWR International
VWR® Sleeve Stoppers
: VWR International
Accessories for Micro-Osmometer, Advanced Instruments
: Advanced Instruments
Lok-Tight™ Contact Plates, Hardy Diagnostics
: Hardy Diagnostics
Potato Dextrose Agar, Hardy Diagnostics
: Hardy Diagnostics
Campy CVA Agar with 5% Sheep Blood, Hardy Diagnostics
: Hardy Diagnostics
EZ-Accu Shot™ Quantitative QC Microorganism Preparation, Microbiologics
: MICROBIOLOGICS, INC.
Tryptic Soy Agar (TSA), Soybean Casein Digest Agar
: Hardy Diagnostics
RECONSTITUTION BUFFER 3 (HCL) 2 ML
: R&D Systems