Eisco QR Code Volumetric Flasks
2000 ml, QR code volumetric flask
Catalog # 470354-614
Supplier: Wards
Supplier #: CH3011QR
Jump to:
Product Details & Documents
Volumetric flasks with industry leading QR code feature.
- Scan the QR code printed directly on the body of the flask to retrieve a PDF of the calibration batch certificate
- Class A Tolerance, conforms to ISO 1042 standards for accuracy
- Autoclavable, with superior chemical durability
- Low temperature gradient - can withstand high temperatures and thermal shocks
Volumetric flasks made from high-quality borosilicate 3.3 glass with a single white graduation marking its capacity. The flask is equipped with a socket fitted with a chemically-resistant polyethylene stopper.
Specifications
- Capacity:2000 ml
- Description:QR code volumetric flask