BSU DNA POLYMERASE L FRAG 2000UNIT 5U/UL
Catalog # MSPP-35872000
Supplier: Intact Genomics
BSU DNA POLYMERASE L FRAG 2000UNIT 5U/UL
Catalog # MSPP-35872000
Supplier: Intact Genomics
Supplier Number: 35872000UNITS
Frequently Bought Together
Immobiline™ DryStrip Gels, Cytiva
Supplier: Cytiva
NAP-BLOCKER™ Non-Animal Protein Blocking Agent, G-Biosciences
Supplier: G-Biosciences
AGAROSE ALFA SELECTOR ST MAGNETIC 2ML
Supplier: NANOTAG BIOTECHNOLOGIES GMBH MS
Deoxynucleotides (dNTPs), G-Biosciences
Supplier: G-Biosciences
ELISA KIT ARG8 VASOPRESSINS
Supplier: ARBOR ASSAYS MS
DNA Elution Buffer, Zymo Research
Supplier: Zymo Research
Blot Boxes, IBI Scientific
Supplier: IBI Scientific
Incubation Trays, Azure Biosystems
Supplier: Azure Biosystems
BREADBOARD ALUMINM M6-TAP 150X600X12.7MM
Supplier: THORLABS INC MS
Pierce™ Protein A/G Magnetic Agarose, Thermo Scientific
Supplier: Invitrogen
IPG Buffers, Cytiva
Supplier: Cytiva
Upright Stainless Steel Freezer Rack for 2" Boxes, 5×4 Configuration
Supplier: Alkali Scientific
OSM (209AA) HUMAN >95% 10UG
Supplier: Genscript
BUFFER LDS SAMPLE LOADING (4X) 10ML
Supplier: G-Biosciences
100-Place Microscope Slide Box
Supplier: Alkali Scientific
RNA RECOVERY BUFFER 10 ML
Supplier: Zymo Research
Western Blot Boxes
Supplier: Alkali Scientific
Electrophoresis Running Buffers, G-Biosciences
Supplier: G-Biosciences
GelBond® Film
Supplier: Lonza