MITAPIVAT-10 MM 1ML IN DMSO SOLID-SOLVNT
Catalog # MSPP-HY-12689
Supplier: 0000031932
MITAPIVAT-10 MM 1ML IN DMSO SOLID-SOLVNT
Catalog # MSPP-HY-12689
Supplier: 0000031932
Supplier Number: HY-12689
Frequently Bought Together
VWR® Razor Blades
Supplier: VWR International
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Reagent alcohol ACS (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
2-Propanol ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Sterile 70% Isopropanol
Supplier: VWR International
VWR® Lead-Free Autoclave Tapes
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Petri Dishes
Supplier: VWR International