TYPE IV COLLAGEN LYOPH POWDER ST HU 5MG
Catalog # MSPP-5022
Supplier: ADVANCED BIOMATRIX, INC. MS
TYPE IV COLLAGEN LYOPH POWDER ST HU 5MG
Catalog # MSPP-5022
Supplier: ADVANCED BIOMATRIX, INC. MS
Supplier Number: 5022-5MG
Frequently Bought Together
Cell Culture Storage Bottles, Polystyrene, Corning
Supplier: Corning
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Corning® Internal Thread Cryogenic Vials, Corning
Supplier: Corning
VWR® Razor Blades
Supplier: VWR International
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
Costar® Multiple Well Cell Culture Plates, Corning
Supplier: Corning
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
Corning® Treated Culture Dishes, Corning
Supplier: Corning
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Nitrile Examination Gloves, Black
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International