Order Entry
United States
Orders LinkContactUsLinkComponent
Sterile Solution Bottles

Sterile Solution Bottles

250 ml, CellPro™ Reservoir bottle, round, Sterile, 24/pack
Catalog # 104028-974
Marketsource
Supplier:   Alkali Scientific
Supplier #:  VR250
Sterile Solution Bottles
Catalog # 104028-974
Supplier:  Alkali Scientific
Marketsource
Sterile Solution Bottles
Sterile Solution Bottles

250 ml, CellPro™ Reservoir bottle, round, Sterile, 24/pack

Price
$138.40
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Store solutions securely with CellPro™ Solution Bottles, individually wrapped and sterile to ensure optimal storage conditions for laboratory reagents.

  • Compatible with CellPro™ Bottle Top Filtration Filters
  • Lightweight yet durable wall construction
  • Wide opening for quick access and non-drip pouring/decanting
  • Non-pyrogenic
  • Polystyrene (PS) bottles are packaged sterile, and individually wrapped
  • Graduated in 25 ml molded increments for accurate filling
  • Single packed sterile bottle and single packed sterile cap
  • Eash case features lot number identification

CellPro™ Solution Bottles offer a sterile and convenient solution for various laboratory applications. Each bottle is individually wrapped to maintain sterility and ensure reliable performance. Designed for versatility, these solution bottles are suitable for storing and dispensing various solutions used in laboratory experiments, research, and clinical settings. With their sterile packaging and robust construction, CellPro™ Solution Bottles provide peace of mind and reliability for any laboratory's needs.

Specifications
  • Sterility:
    Sterile
  • Capacity:
    250 ml
  • Description:
    CellPro™ Reservoir bottle, round
  • Packaging:
    24/pack
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.