Order Entry
United States
Orders LinkContactUsLinkComponent
CellPro™ Flow Cytometry Tubes

CellPro™ Flow Cytometry Tubes

CellPro™ Flow cytometry tubes without caps, bulk econo-pack, 4000 per case, -
Catalog # 104028-366
Marketsource
Supplier:   Alkali Scientific
Supplier #:  CT6420
CellPro™ Flow Cytometry Tubes
Catalog # 104028-366
Supplier:  Alkali Scientific
Marketsource
CellPro™ Flow Cytometry Tubes
CellPro™ Flow Cytometry Tubes

CellPro™ Flow cytometry tubes without caps, bulk econo-pack, 4000 per case, -

Price
$346.09
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Designed for flow cytometry/FACS applications the CellPro™ flow cytometry tubes are made of clear polystyrene and are designed for fluorescence activated cell Sorting/flow cytometry equipment including FACS systems from Becton Dickinson (BD Biosciences). The diameter and wall thickness of the tube's neck is held to very tight tolerances to ensure consistent compatibility with automated instruments.

  • Compatible with FACS (BD) instruments
  • Available with 35 µm strainer cap, standard cap, or without cap
  • Clear polystyrene
  • 5 ml capacity
Specifications
  • Description:
    CellPro™ Flow cytometry tubes without caps, bulk econo-pack
  • Packaging:
    4000 per case
  • Sterile:
    -