Order Entry
United States
Orders LinkContactUsLinkComponent
CellPro™ Flow Cytometry Tubes

CellPro™ Flow Cytometry Tubes

CellPro™ Flow cytometry caps with 35 µm mesh, dual position, for 12×75 mm tubes, 20 Bags of 25, 500 per case, -
Catalog # 104027-370
Marketsource
Supplier #:  CT6409
CellPro™ Flow Cytometry Tubes
Catalog # 104027-370
Supplier:  
Marketsource
CellPro™ Flow Cytometry Tubes
CellPro™ Flow Cytometry Tubes

CellPro™ Flow cytometry caps with 35 µm mesh, dual position, for 12×75 mm tubes, 20 Bags of 25, 500 per case, -

Discontinued
Product Details & Documents

Designed for flow cytometry/FACS applications the CellPro™ flow cytometry tubes are made of clear polystyrene and are designed for fluorescence activated cell Sorting/flow cytometry equipment including FACS systems from Becton Dickinson (BD Biosciences). The diameter and wall thickness of the tube's neck is held to very tight tolerances to ensure consistent compatibility with automated instruments.

  • Compatible with FACS (BD) instruments
  • Available with 35 µm strainer cap, standard cap, or without cap
  • Clear polystyrene
  • 5 ml capacity
Specifications
  • Description:
    CellPro™ Flow cytometry caps with 35 µm mesh, dual position, for 12×75 mm tubes
  • Packaging:
    20 Bags of 25, 500 per case
  • Sterile:
    -
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.