Orders LinkContactUsLinkComponent
Bulk Pipet Tips
Bulk Pipet Tips
50 - 1250 µl, -, 1000/pack, bulk bagged
Catalog # 104028-140
Marketsource
Supplier:   Alkali Scientific
Supplier #:  BT1250
Bulk Pipet Tips
Catalog # 104028-140
Supplier:  Alkali Scientific
Marketsource
Bulk Pipet Tips
Bulk Pipet Tips
50 - 1250 µl, -, 1000/pack, bulk bagged
Price
$37.32
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Ensure precision and efficiency with Alkali Scientific™ bulk pipet tips.
  • Fine point design
  • Certified Rnase, Dnase and pyrogen free
  • Universal fit
  • Ultra clear polypropylene
  • Withstands organic solvents
Bulk pipet tips are indispensable laboratory consumables designed for precise and accurate liquid handling in various scientific applications. Typically made from high-quality polypropylene or similar materials, these tips offer excellent chemical resistance and minimize sample contamination. Packaged in large quantities, often in bags, racks, or boxes containing hundreds or thousands of tips per pack, bulk pipet tips ensure an ample supply for routine laboratory tasks and high-throughput experiments, reducing the frequency of tip changes and workflow interruptions.
Available in various volume capacities and compatible with both manual and automated pipetting systems, these tips provide reliability, consistency, and cost-effectiveness in laboratories engaged in molecular biology, biochemistry, cell culture, microbiology and other scientific disciplines requiring accurate liquid dispensing.
Specifications
  • Volume:
    50 - 1250 µl
  • Packed:
    1000/pack, bulk bagged
  • Sterile:
    -