Orders LinkContactUsLinkComponent
CellPro™ Neutralizing Solutions
CellPro™ Neutralizing Solutions
CellPro™ Neutralizing solution, 4 L cubitainer, Cubitainer®, 4 L
Catalog # 104028-868
Marketsource
Supplier:   Alkali Scientific
Supplier #:  DS0215-4L
CellPro™ Neutralizing Solutions
Catalog # 104028-868
Supplier:  Alkali Scientific
Marketsource
CAS Number:  
CellPro™ Neutralizing Solutions
CellPro™ Neutralizing Solutions
CellPro™ Neutralizing solution, 4 L cubitainer, Cubitainer®, 4 L
CAS Number:  
Price
$193.78
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
CellPro™ Neutralizing Solution is an essential reagent in molecular biology and biochemical experiments, particularly in protocols involving nucleic acid hybridization and blotting techniques.
  • Sterile
  • Filtered
This solution is specifically formulated to neutralize the effects of strong bases or acids used during the denaturation process. For example, after treating samples with denaturing solutions such as 0.5 M NaOH with 1.5 M NaCl, the CellPro™ Neutralizing Solution effectively restores the pH balance, creating an optimal environment for subsequent experimental steps.
Caution: UN No. 3248
Specifications
Specifications
  • Pack type:
    Cubitainer®
  • Description:
    CellPro™ Neutralizing solution, 4 L cubitainer
  • Size:
    4 L
  • Cat. No.:
    104028-868
  • CAS No.:
    367-93-1