Order Entry
United States
Orders LinkContactUsLinkComponent
CellPro™ Borate Buffer, Sterile, Filtered

CellPro™ Borate Buffer, Sterile, Filtered

CellPro™ Borate buffer, 0.5 M, 500 ml, 8.5
Catalog # 104026-700
Marketsource
Supplier:   Alkali Scientific
Supplier #:  DB0217
CellPro™ Borate Buffer, Sterile, Filtered
Catalog # 104026-700
Supplier:  Alkali Scientific
Marketsource
CAS Number:  
CellPro™ Borate Buffer, Sterile, Filtered
CellPro™ Borate Buffer, Sterile, Filtered

CellPro™ Borate buffer, 0.5 M, 500 ml, 8.5

CAS Number:  
Price
$103.78
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Optimize your experiments with CellPro™ borate buffer, offering sterile and filtered solutions for consistent performance.

  • Colorless
  • Sterile
  • Research Grade

CellPro™ Borate buffer delivers a sterile and filtered solution essential for a wide range of laboratory procedures. Crafted with precision, this buffer solution ensures purity and reliability in scientific investigations, meeting the exacting standards of research and experimentation. The sterile nature of the buffer minimizes the risk of contamination, making it suitable for various biochemical assays, enzyme studies, and molecular biology applications. With its filtered formulation, CellPro™ borate buffer guarantees clarity and consistency in experimental outcomes, providing researchers with confidence in their results. Whether employed for protein analysis, nucleic acid electrophoresis, or other laboratory protocols, this buffer solution offers a dependable and standardized option for diverse scientific needs.

Specifications

Specifications

  • Description:
    CellPro™ Borate buffer, 0.5 M
  • Size:
    500 ml
  • pH:
    8.5

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.