CellPro™ ACK Lysing Buffer, Sterile
CellPro™ ACK lysing buffer, 1 L
Catalog # 104026-770
Marketsource
Supplier: Alkali Scientific
: DB0309
CellPro™ ACK Lysing Buffer, Sterile
Catalog # 104026-770
Supplier: Alkali Scientific
Marketsource
CAS Number:
Jump to:
Product Details & Documents
Efficiently lyse red blood cells with CellPro™ ACK Lysing Buffer, ensuring reliability and sterility in your experiments.
- For the lysis of red blood cells in samples containing white blood cells
- Sterile
- Filtered
CellPro™ ACK Lysing Buffer is a sterile solution designed for efficient red blood cell lysis in biological samples. This specialized buffer ensures the gentle and thorough removal of erythrocytes, facilitating downstream analysis in flow cytometry, cell sorting, and other hematological applications.
With its sterile formulation, CellPro™ ACK Lysing Buffer maintains sample integrity and minimizes the risk of contamination, ensuring reliable and reproducible results. Streamline your cell analysis workflows with the high-quality performance and convenience of CellPro™ ACK Lysing Buffer.
Specifications
Specifications
- Description:CellPro™ ACK lysing buffer
- Size:1 L