CellPro™ Denaturing Solution, Alkali Scientific
CellPro™ Denaturing solution, Plastic bottle, 1 L, 12/case
104028-736
Marketsource
: DB031-CS
Presenting CellPro™ Denaturing Solution, meticulously crafted by to exacting standards for your molecular biology endeavors.
- Designed for use in Southern and northern blotting, and in situ hybridization procedures
- Sterile
- Filtered
This sterile filtered denaturing solution offers unparalleled purity and reliability, ensuring consistent performance in your experiments. Whether you're conducting nucleic acid denaturation, DNA sequencing, or other molecular biology applications, CellPro™ Denaturing solution provides the optimal environment for accurate and reproducible results. Trust in CellPro™ Denaturing solution to meet your research needs with precision, allowing you to pursue scientific inquiry with confidence and clarity.
- Pack type:Plastic bottle
- Description:CellPro™ Denaturing solution
- Size:1 L, 12/case
- Cat. No.:104028-736
- CAS No.:1310-73-2;7647-14-5