CellPro™ Neutralizing Solutions
CellPro™ Neutralizing Solution is an essential reagent in molecular biology and biochemical experiments, particularly in protocols involving nucleic acid hybridization and blotting techniques.
- Sterile
- Filtered
This solution is specifically formulated to neutralize the effects of strong bases or acids used during the denaturation process. For example, after treating samples with denaturing solutions such as 0.5 M NaOH with 1.5 M NaCl, the CellPro™ Neutralizing Solution effectively restores the pH balance, creating an optimal environment for subsequent experimental steps.
: UN No. 3248
- Pack type:Plastic bottle
- Description:CellPro™ Neutralizing Solution
- Size:1 L
- Cat. No.:104027-500
- CAS No.:77-86-1;7558-80-7;7558-79-4